Bouffant Caps, Veltek
Bouffant Caps, Veltek
Supplier:   Veltek Associates
Bouffant Caps, Veltek
Supplier:  Veltek Associates
Certificates
Bouffant Caps, Veltek
2 Options Available Below
Product Details & Documents
These bouffant hats are designed and manufactured in a pleated design that provides full coverage.
  • Lightweight, cool, breathable
  • Latex free
For use in cleanroom settings.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 75795-412
  • 75795-358
  • Fabric:
  • PP
  • PP
  • Garment application:
  • Medical|Clinical|Cleanroom|Laboratory|Pharmaceutical
  • Medical|Clinical|Cleanroom|Laboratory|Pharmaceutical
Spécifications
Product Family Options
Informations sur le produit
  • Color
    Ø
    Availability
    Price
  • Blue
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $199.00
    $199.00
    Case of 1,000
       $199.00
    Supplier Number: 150-BF-NS-1530
    Spécifications:
    More
    Specifications
    Cat. No.
    75795-412
    Fabric
    PP
    Garment application
    Medical|Clinical|Cleanroom|Laboratory|Pharmaceutical
  • Blue
    23.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $218.00
    $218.00
    Case of 1,000
       $218.00
    Supplier Number: 150-BF-NS-1531
    Spécifications:
    More
    Specifications
    Cat. No.
    75795-358
    Fabric
    PP
    Garment application
    Medical|Clinical|Cleanroom|Laboratory|Pharmaceutical