One-Piece Shovel with 14" Blade, Remco
Supplier: Remco Products
Certificates
About this item
Remco one-piece polypropylene shovels are tough, lightweight and hygienic.
- Molded from FDA-compliant polypropylene
- Autoclavable at 250 °F/121 °C
This is a workhorse for moving bulk materials and an integral part of any professional cross–contamination containment, food safety or HACCP program. This shovel is popular for meat, dairy, seafood, and poultry processing, as well as commercial fishing and other food handling applications.
Delivery: Dropshipping available.
Frequently Bought Together