Basic Cotton Coveralls, Dickies®
Supplier: Dickies
À propos de cet article
Dickies® style 48300, long sleeve.
- Bi-swing back
- Elastic waist inserts
- Heavy-duty brass two-way zipper
- Concealed neck and waist snap
- Handy ruler pocket
- Two large back pockets
- 6 oz. twill, 100% cotton
21 Options Available Below
Product Details & Documents
Dickies® style 48300, long sleeve.
- Bi-swing back
- Elastic waist inserts
- Heavy-duty brass two-way zipper
- Concealed neck and waist snap
- Handy ruler pocket
- Two large back pockets
- 6 oz. twill, 100% cotton
Specifications for Product Family
Spécifications
- Catalog No:
- 76168-578
- 76168-576
- 76168-580
- 76168-128
- 76168-560
- 76168-562
- 76168-564
- 76168-566
- 76168-568
- 76165-802
- 76165-798
- 76165-800
- 76165-794
- 76165-796
- 76165-804
- 76165-806
- 76168-130
- 76165-786
- 76165-788
- 76165-790
- 76165-792
- 76168-570
- 76168-572
- 76168-574
- Model:
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- 48300
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Gender:
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Twill, 100% cotton
- Thickness:
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
- 6 oz.
Spécifications
Product Family Options
Informations sur le produit
- ColorTaille de confectionCoupe du tenueAvailabilityPrice
- Supplier Number: 48300DNSRGSpécifications:
More
Specifications
Cat. No.76168-578Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300DNMTLSpécifications:
More
Specifications
Cat. No.76168-576Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300DNXLRGSpécifications:
More
Specifications
Cat. No.76168-580Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300DNXLTLSpécifications:
More
Specifications
Cat. No.76168-128Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300DN2XRGSpécifications:
More
Specifications
Cat. No.76168-560Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300DN2XTLSpécifications:
More
Specifications
Cat. No.76168-562Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300DN3XRGSpécifications:
More
Specifications
Cat. No.76168-564Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300DN3XTLSpécifications:
More
Specifications
Cat. No.76168-566Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300DN4XRGSpécifications:
More
Specifications
Cat. No.76168-568Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KHSRGSpécifications:
More
Specifications
Cat. No.76165-802Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KHMRGSpécifications:
More
Specifications
Cat. No.76165-798Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KHMTLSpécifications:
More
Specifications
Cat. No.76165-800Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KHLRGSpécifications:
More
Specifications
Cat. No.76165-794Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KHLTLSpécifications:
More
Specifications
Cat. No.76165-796Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KHXLRGSpécifications:
More
Specifications
Cat. No.76165-804Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KHXLTLSpécifications:
More
Specifications
Cat. No.76165-806Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KH2XRGSpécifications:
More
Specifications
Cat. No.76168-130Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KH2XTLSpécifications:
More
Specifications
Cat. No.76165-786Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KH3XRGSpécifications:
More
Specifications
Cat. No.76165-788Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KH3XTLSpécifications:
More
Specifications
Cat. No.76165-790Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Supplier Number: 48300KH4XRGSpécifications:
More
Specifications
Cat. No.76165-792Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz. - Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.76168-570ModelGarment applicationGenderSleeve or leg lengthClosure typeFabricTwill, 100% cottonThickness6 oz. - Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.76168-572ModelGarment applicationGenderSleeve or leg lengthClosure typeFabricTwill, 100% cottonThickness6 oz. - Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.76168-560Model48300Garment applicationGeneral PurposeGenderFor MenSleeve or leg lengthLongClosure typeZip closureFabricTwill, 100% cottonThickness6 oz.