Versaflo™ High Efficiency Filter, 3M™
High Efficiency Filter
10152-834
: 3M
: 7100019288
TR-6710N is a High Efficiency (HE) particulate filter for use with TR-600 Series PAPRs. The filter attaches directly to the motor/blower. 3M™ recommends the use of the TR-6700FC filter cover to help protect the filter from physical damage. The TR-6700FC filter cover is also used to hold the TR-6600 prefilter or TR-662 spark arrestor in place.
- High efficiency filter
- Bulk packaging for convenience
- Simple to install
- Désignation:High Efficiency Filter