About this item
Grind the toughest samples at cryogenic temperatures.
- Reach cryogenic temperatures in minutes
- Dual grinding and cooling chambers available
- User-friendly touch screen controls
- 20 user-defined programs for easy quick recall
- Enhanced safety features
The Freezer/Mill® programmable cryogenic grinder pulverizes through the toughest samples at cryogenic temperatures. Samples are placed in a vial and immersed in a self-contained insulated liquid nitrogen (LN2) tub, reaching cryogenic temperatures in minutes, where they are then pulverized with a magnetically driven impactor. The closed grinding vial impactor prevents sample cross-contamination. The optional dual grinding and cooling chambers allow you to pre-cool and grind two sets of samples simultaneously.
Grind temperature sensitive samples while maintaining constant temperature, as the sample never leaves the enclosed liquid nitrogen bath. Enhanced safety features include a liquid nitrogen sensor and lid interlock. If the lid is opened while in process, grinding will stop. Auto-fill system is available for automated liquid nitrogen filling.
The large, high-resolution touch screen controls all parameters of the run. Store up to 20 user-defined grinding programs for quick and easy recall. The user-friendly interface allows you to view training videos, user manual, and run history, which can be transferred to a computer using a thumb drive.
Specifications
- Catalog No:
- 76182-008
- 76182-006
- 76182-012
- 76182-010
- 76182-014
- 76182-004
- 76181-978
- 76181-976
- Dimensions:
- 52×55×46 cm (20.5×21.5×18")
- 52×55×46 cm (20.5×21.5×18")
- 50.8×55×64.8 cm (20×21.5×25.5")
- 52×55×46 cm (20.5×21.5×18")
- 50.8×55×64.8 cm (20×21.5×25.5")
- 52×55×46 cm (20.5×21.5×18")
- 48×27×32 cm (19×10.5×12.75")
- 48×27×32 cm (19×10.5×12.75")