SP Wilmad-LabGlass Bend Adapters with Hose Connection and Standard Taper Joint, SP Industries
14206-312
- ST Joint:24/40
- Environmentally Preferable:
- Cat. no.:14206-312
- Hose Connection O.D.:⅜"
- Supplier No.:LG-1702-102
These borosilicate glass adapters have a 90° bend to hose connector at top and a standard taper inner joint at bottom.