Vikan® Condensation Squeegee, 16", Remco Products
Fournisseur: Remco Products
Certificats
À propos de cet article
Squeegee features drain holes for effective removal of condensation from ceilings and pipes.
- Made with FDA compliant materials
- Autoclavable at 121 °C (250 °F)
Squeegee with drain holes allows effective removal of condensation from ceilings and pipes. Collection of condensation prevents moisture from dripping into food product or on costly machinery.
Delivery: Dropshipping available.
Frequently Bought Together