Airfree® Reaction Flasks, Round Bottom, Schlenk, Chemglass
These round bottom single neck flask having a standard taper outer joint.
Available with a choice of a 2 mm high vacuum glass stopcock having a S.S. constant pressure retaining device or a 0 to 4 mm Chem-Cap® Airfree® inlet valve.
Frequently Bought Together