ContactUsLinkComponent
Forensic Drug Testing: A Simulated Immunoassay
  470237-798
undefined
Forensic Drug Testing: A Simulated Immunoassay
  470237-798
 :  IS9023
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Forensic Drug Testing
  • No. of Lab Groups:
    15
  • Cat. no.:
    470237-798
  • Includes:
    Teachers Guide and Student Copymaster
Real-Life Forensic Urine Analysis.
  • Perfect for middle school students
  • Aligned to NGSS LS1.A, PS1.A and PS1. B
Drug testing comes in two forms, presumptive and confirmatory. Because of the cost associated with confirmatory testing, a presumptive test is often employed first. While there are various forms of presumptive drug testing, two of the most common techniques are immunoassay testing and thin-layer chromatography. In this activity, students will perform a simulated immunoassay test on several urine samples for the detection of the cocaine metabolite benzoylecgonine. All urine samples and testing reagents are simulated to avoid the potential hazards of actual biological specimens while still providing realistic results.