Exploring Meteorology | Avantor
Exploring Meteorology
Exploring Meteorology, Teachers Guide and Student Copymaster, 15
Catalog # 470237-788
Supplier:   Ward's Science
Fournisseur #:  IS8706
Exploring Meteorology
Catalog # 470237-788
Supplier:  Ward's Science
Exploring Meteorology
Exploring Meteorology
Exploring Meteorology, Teachers Guide and Student Copymaster, 15
Price
$109.00
Log in pour la disponibilité
Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Learn how to meteorologists predict the weather.
  • Perfect for middle and high school students
  • Aligned to NGSS ESS2.C and ESS2.D
Weather impacts everyone, everyday in some way and the meteorologist that study weather patterns are our best source for information. This experiment provides students with the experience of being a meteorologist. It includes studies of pressure changes and creating their own barometer. Studies of wind speed and creating their own anemometer and, the skills to confidently read a weather map. *Bonus* this experiment includes a demonstration of how clouds form by allowing the instructor to form a vapor cloud. Experiment includes enough material for 15 groups of 2 to perform the tasks. The demonstration portion has enough material to perform the demonstration 5 times.
Spécifications
  • Désignation:
    Exploring Meteorology
  • No. of Lab Groups:
    15
  • Includes:
    Teachers Guide and Student Copymaster