Minerals Model Set
Minerals Model Set
Catalog # 470232-074
Supplier: Mega Molecules
Fournisseur #: GC430
Jump to:
Product Details & Documents
Hands on mineral crystal structures.
- Includes 1 activity
- Perfect for high school and college students
Build the most abundant silicate minerals including feldspar, quartz, olivine, pyroxenes, amphiboles, micas, and clay minerals. Learn how models of silicate minerals are related to unit cell and crystal lattice structures. This model set has enough parts to construct all six types of silicate minerals at one time allowing students to compare nesosilicates, sorosilicates, cyclosilicates, inosilicates, phyllosilicates, and tectosilicates.
Spécifications
- Désignation:Minerals Model Set