Casein Enzyme Hydrolysate, Type - II, HiMedia
Catalog # CA61001-574
Supplier: HiMedia
CAS Number:
Specifications
- Description:Casein Enzyme Hydrolysate, Type - II
- Size:500 g
- Cat. no.:CA61001-574
- Supplier No.:RM028-500G
Specifications
About this item
This is equivalent to Casitone and is used in antibiotic assay media.