Caps for Mobile Phase Bottles, Microsolv
Caps for Mobile Phase Bottles, Microsolv
Caps for Mobile Phase Bottles, Microsolv
Caps for Mobile Phase Bottles, Microsolv
These caps are designed for use on HPLC mobile phase reservoir and storage bottles with GL 45 thread.
  • Made up of polypropylene
Product Family Options
  • Matière
    Color
    Epaisseur
  • PP
    Blue
    GL-45
    Pack of 10
    Pack of 10 - $98.68
    $98.68
    Pack of 10
       $98.68
     : 58807-B45
  • PP
    Green
    GL-45
    Pack of 10
    Pack of 10 - $98.68
    $98.68
    Pack of 10
       $98.68
     : 58807-G45
  • PP
    Purple
    GL-45
    Pack of 10
    Pack of 10 - $98.68
    $98.68
    Pack of 10
       $98.68
     : 58807-P45
  • PP
    Yellow
    GL-45
    Pack of 10
    Pack of 10 - $98.68
    $98.68
    Pack of 10
       $98.68
     : 58807-Y45