ContactUsLinkComponent
DA-7645® Process and Research Cleaners
Catalogue # 77531-102
Fournisseur:  Steris
New Product
DA-7645® Process and Research Cleaners
Catalogue # 77531-102
Fournisseur:  Steris
Supplier Number:  1165D3
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1136 L (300 gal.)
  • Cat. no.:
    77531-102
  • Packaging:
    Plastic tote
Spécifications
À propos de cet article
DA-7645® Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms in manual cleaning applications.
  • Efficacy - Mild pH formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Productivity - Delivers efficiency to manual cleaning operations using a blend of surfactants, solvents, and other performance-enhancing ingredients
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
DA-7645® Process and research cleaner is specially formulated to enhance manual cleaning with enhanced substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.
Ordering information: .