Whatman™ Moisture Test Paper, Whatman products (Cytiva)
CA76205-426
: Cytiva
- Ø:90 mm
- Max. operating temperature:550 °C
- Cat. no.:CA76205-426
These borosilicate glass papers are designed for use when drying samples during moisture assessment.
Max. recommended temperature: 550 °C