ContactUsLinkComponent
Whatman™ Moisture Test Paper, Whatman products (Cytiva)
  CA76205-426
 :  Cytiva
undefined
Whatman™ Moisture Test Paper, Whatman products (Cytiva)
  CA76205-426
 :  Cytiva
 :  5401-090E
  • Ø:
    90 mm
  • Max. operating temperature:
    550 °C
  • Cat. no.:
    CA76205-426
These borosilicate glass papers are designed for use when drying samples during moisture assessment.
Max. recommended temperature: 550 °C