SP Wilmad-LabGlass Precision Grade Graduated Cylinders with Pour-Out, SP Industries
Having a great level of precision and conforming to ASTM E1272 Style 1 Class A, these borosilicate glass graduated cylinders feature a reinforced bead and pouring lip.
Calibrated “to deliver” at 20 °C and suitable for general laboratory use.
Frequently Bought Together