Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Length:11.4 cm (4½")
- Material:Stainless steel
- Tip Configuration:2 x 3 Prongs, Blunt
- Cat. no.:76192-462
The Econo™ Weitlaner Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The Econo™ Weitlaner Retractor has curved shanks that lead to blades that have between two and six, sharp or blunt, outward curving prongs. The prongs interdigitate when the instrument is closed. The Weitlaner retractor is a popular instrument, most commonly used in basic plastic surgery, large bone and joint procedures, and mastoid surgeries. It is also useful with small, deep incisions and soft tissue dissection at a superficial level. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.