Archer Splinter Forceps, OR Grade, Sklar®
Product Details & Documents
Sklar® Archer Splinter Forceps are used for removing foreign objects from the surface of the skin.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Sklars® Archer Splinter Forceps have a straight, sharp, serrated tip with finger ring handles for better grip.
Spécifications
- Longueur:5.5"
- Matière:Stainless steel
- Curvature:Straight
- Tip Style:Serrated