Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Length:18.4 cm (7¼")
- Material:Stainless steel
- Curvature:Slightly Curved
- Cat. no.:76192-636
Specifications
About this item
The Econo™ Freer Elevator is an orthopedic instrument that can be used as a multipurpose tool in many procedures.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The Econo™ Freer Elevator is primarily used for blunt debulking and lifting periosteum from bones in confined areas. This double-ended instrument has a rounded handle with curved, tear-drop tips, one sharp and one blunt. Econo™ line instruments are floorgrade, disposable instruments sold in bulk quantities.