Biox Microtube Rack
Microtube Rack, Assorted, 80, PP
Supplier: TWD Scientific
Supplier #: BX90-8009
Jump to:
Product Details & Documents
Microcentrifuge Tube Racks are made of a high-strength polypropylene that is chemically resistant to alcohols and mild organic solvents. The molded 5×16, 80-Well array is designed for easy tube identification.
- Polypropylene
- Easy Mark writing area
- Equipped with side handles
- Assorted colors (red, blue, purple, green and yellow)
Microtube racks are designed for use with 1.5 or 2.0 mL microtubes. Racks feature a molded numeric grid for quick identification purposes.
Documents
Specifications
- Nb de places:80
- Color:Assorted
- Désignation:Microtube Rack
- Matière:PP