DuPont™ ProClean® Frocks, Front Zipper Closure
Supplier: DuPont
Certificates
About this item
ProClean® offers improved non-hazardous light liquid splash protection.
- Serged seams
- Bound neck
- Set sleeve design
- Covered elastic wrists
- Front zipper closure
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Covered elastic openings at wrists allows for a tighter fit. Bound neck ensures lower particle shedding. Frocks provide body-length jacket design with front zipper closure for easy donning and doffing.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Specifications
- Catalog No:
- CA13452-546
- CA13452-550
- CA89129-704
- CA13452-544
- CA13452-552
- CA13452-548
- CA13452-554
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory