Ward's® Sterile Cell Strainers
Ward's® Sterile Cell Strainers
Supplier:  MTC BIO, INC.
Certificates
Ward's® Sterile Cell Strainers
Ward's® Sterile Cell Strainers
6 Options Available Below
Product Details & Documents
Ward's® Sterile Cell Strainers and Cell-Straining Kits provide all the necessary parts for filtration of cell suspensions.
  • Available as Kits (strainer, 50 ml tube, and cap) or as strainers alone
  • Easy to open under the hood, reducing contamination risk
Cell Straining Kits: Eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps). The individual sterile packets mean users are no longer forced to expose entire bags of 50 ml tubes and corresponding caps in order to perform one filtration. Instead they can open a single packet directly under the hood.
Cell Strainers: If you don't need the accompanying tube that comes with the Cell Straining Kit, you can purchase 40 μm, 70 μm, or 100 μm cell strainers by themselves.
Product Family Options
Cell Strainer and Tube Kits
Informations sur le produit
  • Désignation
    Taille de pore
    Color
    Availability
    Price
  • Discontinued
  • Discontinued
  • Large-Mesh Strainer Kit
    100 μm
    Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $534.12
    $534.12
    Pack of 50
       $534.12
Cell Strainers Only
Informations sur le produit
  • Désignation
    Taille de pore
    Color
    Availability
    Price
  • Small-Mesh
    40 μm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $263.25
    $263.25
    Pack of 50
       $263.25
    Supplier Number: C4040_WARDS
  • Medium-Mesh
    70 μm
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $263.25
    $263.25
    Pack of 50
       $263.25
    Supplier Number: C4070_WARDS
  • Large-Mesh
    100 μm
    Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $244.90
    $244.90
    Pack of 50
       $244.90
    Supplier Number: C4100_WARDS
Optional Items
Informations sur le produit
  • Désignation
    Taille de pore
    Color
    Availability
    Price
  • Adapter for 5 and 15 ml Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $97.95
    $97.95
    Pack of 25
       $97.95
    Supplier Number: C4015_WARDS
  • Dual-Sided Pestle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $105.30
    $105.30
    Pack of 50
       $105.30
    Supplier Number: C4010_WARDS