Miracle-Gro | Avantor
Miracle-Gro
Miracle-Gro
Catalog # 470222-128
Supplier:   ARETT SALES CORP
Fournisseur #:  S70-2001123
Miracle-Gro
Catalog # 470222-128
Supplier:  ARETT SALES CORP
Miracle-Gro
Miracle-Gro
Miracle-Gro
Price
$30.20
Log in pour la disponibilité
Each Case of 6
Product Details & Documents
A Great General Purpose Fertilizer!
  • Water soluble
  • For all plants
Water soluble all purpose plant food 24-8-16 is great for all types of plants, vegetables, trees, shrubs and houseplants. Works instantly for quick, beautiful results. Feeds through leaves and roots. Use in either the Miracle-Gro Garden Feeder or watering cans. Guaranteed not to burn when used as directed.
Spécifications
  • Désignation:
    Miracle-Gro