Carson eFLEX Digital Microscope
eFlex, MM-840
Catalog # 470235-858
Supplier: CARSON OPTICAL
Fournisseur #: MM-840
Jump to:
Product Details & Documents
The eFlex from Carson is a powerful digital microscope that displays the magnified image right on your computer screen.
- 1600 x 1200 max picture resolution
- 640 x 480 max video resolution
The impressive 75x/300x effective magnification (on 21 inch monitor) allows you to see details of ordinary objects that you never knew existed.
Capture an image to keep with the click of a button. Captures close-focus video. The included flexible neck stand with suction cup adapter attached to any smooth surface, or included graduated base with ease.
Caution: Does not work with tablets.
Spécifications
- Modèle:MM-840
- Désignation:eFlex
- Computer requirements:Compatible with Windows 10/8/8.1/7/Vista/XP SP2+ and all Mac OS. A USB 2.0/3.0
- Cat. No.:470235-858