ContactUsLinkComponent
Carson HookUpz Universal Smartphone Binocular Adapter
Carson HookUpz Universal Smartphone Binocular Adapter
Catalog # 470235-860
Supplier:  CARSON OPTICAL
undefined
Carson HookUpz Universal Smartphone Binocular Adapter
Catalog # 470235-860
Supplier:  CARSON OPTICAL
Supplier Number:  IB-700
Specifications
  • Description:
    Hookupz Smartphone Bino Adapter IB-700
  • Cat. no.:
    470235-860
Specifications
About this item
The HookUpz Universal Smartphone Binocular Adapter from Carson easily connects your smartphone to most full sized binoculars.
  • Objective Lens Diameter: 32 to 50 mm
Use your built-in camera application on your smartphone to take pictures and record video through the lenses of your binoculars.
Fits all phones. For specific information on your model, visit: www.carson.com/multicam.