Carson Helping Hands Magnifier
Helping Hands GN-88, 2×
Catalog # 470235-854
Supplier: CARSON OPTICAL
Fournisseur #: GN-88
Jump to:
Product Details & Documents
The HelpingHands magnifier from Carson is a hand-held or hands-free, fully adjustable LED lighted soldering and hobby magnifier.
- Magnification: 2× power
It contains a distortion-free acrylic lens (4.3 inch lens diameter).
Spécifications
- Grossissement:2×
- Désignation:Helping Hands GN-88