SEM Conductive Tapes, Electron Microscopy Sciences
À propos de cet article
We now offer a new line of conductive tapes, that are electrically conductive and have a clean background. They were developed especially for scanning electron microscopy.
15 Options Available Below
Product Details & Documents
We now offer a new line of conductive tapes, that are electrically conductive and have a clean background. They were developed especially for scanning electron microscopy.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 77810
- Supplier Number: 77817-05
- Copper/Nickel TapeEach$507.47Pack of 5$414.23Supplier Number: 77811
- Double Sided Carbon TapeEach$524.60Pack of 5$420.03Supplier Number: 77817
- Supplier Number: 77816
- Supplier Number: 77813
- Supplier Number: 77817-12
- Aluminum/Nickel TapeEach$412.20Pack of 5$333.30Supplier Number: 77814
- Supplier Number: 77817-20
- Supplier Number: 77817-50
- Supplier Number: 77817-05-AL
- Supplier Number: 77817-08-AL
- Supplier Number: 77817-12-AL
- Supplier Number: 77817-20-AL
- Supplier Number: 77817-50-AL