Sterile SatPax® 1000 Pre-Wetted Cellulose/Polyester Nonwoven Cleanroom Wipes, Berkshire
Catalogue # 76204-772
Fournisseur: Berkshire
Spécifications
- Description:Sterile SatPax® 1000 Wipes, C-Fold
- Cat. no.:76204-772
- Packaging:30 Wipes/Pack, 24 Packs/Case (Stacked)
- W×L:22.9×22.9 cm (9×9)"
Spécifications
À propos de cet article
Gamma irradiated and sterile validated to a 10–6 Sterility Assurance Level per AAMI guidelines.
- Low extractables and fiber and particle counts
- Smooth, highly absorbent, and durable
- Low extractable levels and particle counts
- High saturation level is ideal for removing cleaning and disinfecting residues in sterile environments
Sterile SatPax® 1000 combines Durx® 670 nonwoven wipers composed of a hydroentangled nonwoven blend of 55% cellulose and 45% polyester with a high saturation level of 70% IPA and 30% DI water. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles, and inconsistent wetting and cleaning associated with wetting a dry wiper. Making the product ideal for cleaning critical sterile processing environments.
Certifications: Designed for use in ISO class 5 and higher cleanroom environments. Cleanroom Class 100 – 100,000.