- Description:Robolink CoDrone EDU
- Dimensions:124×138×35 mm
- Cat. no.:470235-890
- Weight:57 g
A programmable drone, made for learning in the classroom.
- Perfect for ages 12+
- Code in Blockly or Python
- 60 min battery charge
Introducing the CoDrone EDU, the perfect drone for the classroom. With years of experience using drones for learning, we've designed a drone that puts learning first. It provides open access to code and sensor data for talking about STEM topics, and enough durability and repairability to withstand the rigors of classroom use.
Code it in Python or start from the basics with block-based coding. Unique to this drone are its multiple sensors - gyroscope, barometer, color, front and bottom range sensors, and more - all usable in code. With a durable and safe frame, programmable lights, and access to its 7 sensors, it's an excellent learning tool and a great way to learn coding and engineering skills.