Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
Product Family Options
  • Matière
    Septum
    Taille du bouchon
    Cap Color
    Cap Type
  • PP
    Red Orange Natural Rubber/ Transparent TFE
    13 - 425 mm
    Black
    Screw cap
    Pack of 100
    Pack of 100 - $94.62
    $94.62
    Pack of 100
       $94.62
     : 9504S-80C-BK
  • PP
    Silicone Rubber / PTFE Pre-Slit
    13 - 425 mm
    Black
    Screw cap
    Pack of 100
    Pack of 100 - $66.87
    $66.87
    Pack of 100
       $66.87
     : 9504S-35C-BLK
  • PP
    Silicone Rubber / PTFE Pre-Slit
    13 - 425 mm
    White
    Screw cap
    Pack of 100
    Pack of 100 - $73.04
    $73.04
    Pack of 100
       $73.04
     : 9504S-30C-W
  • PP
    Silicone Rubber / PTFE Pre-Slit
    13 - 425 mm
    White
    Screw cap
    Case of 1,000
    Case of 1,000 - $630.23
    $630.23
    Case of 1,000
       $630.23
     : 9504S-30C-W-K