Taq2000 DNA Polymerase, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Certificates
About this item
Highly purified, recombinant Taq DNA Polymerase cloned from thermophilic eubacteria, Thermus aquaticus.
- Ultrapure cloned Taq DNA polymerase
- Virtually eliminates background artifacts
- Minimizes smearing in long PCR
- Most thermostable Taq DNA polymerase
Taq2000 DNA Polymerase is a highly purified, recombinant Taq DNA polymerase cloned from the thermophilic eubacteria, Thermus aquaticus.
Caution: For research use only. Not for use in diagnostic procedures.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together