Human/Mouse Reference RNA, Agilent Technologies
Total RNA which has been extracted from cell lines representing different human tissues, pooled (10 human) or pooled (11 mouse), and treated to remove DNA to undetectable levels.
- Greater consistency than RNA references derived from tissue sources
- Maximum representation of low, medium, and high abundant gene transcripts
- Manufactured at industrial scale lot sizes and stringently quality controlled for minimized lot-to-lot variation
- No detectable levels of genomic DNA
Typical uses include optimization of primer/probe sets to ensure no cross reactivity, as a standard to relatively quantitate unknowns to an unchanging reference for immediate validation without wasting precious RNA, or to compare QPCR efficiencies across labs/platforms/experiments.
: For research use only. Not for use in diagnostic procedures.
Frequently Bought Together