Tourniquets
Supplier: Dukal
Jump to:
À propos de cet article
Latex-free tourniquets are ideal for constricting or compression.
- Feels like latex
- Hypoallergenic
- Superior strength
- Convenient packaging designed for kit assembly and resale
- Not made with natural rubber latex
- Weight: 15.29 lbs.
Made of synthetic polyisoprene.
3 Options Available Below
Product Details & Documents
Latex-free tourniquets are ideal for constricting or compression.
- Feels like latex
- Hypoallergenic
- Superior strength
- Convenient packaging designed for kit assembly and resale
- Not made with natural rubber latex
- Weight: 15.29 lbs.
Made of synthetic polyisoprene.
Documents
Certificates
Recherche par numéro de lotProduct Family Options
Informations sur le produit
- DésignationPackagingAvailabilityPrice
- Non sterile tourniquet, blue, flat, 1×18"100/box, 10 boxes/CaseLog in pour la disponibilitéCase of 1,000$225.39Supplier Number: 4100
- Non sterile tourniquet, orange, flat, 1×18"100/box, 10 boxes/CaseLog in pour la disponibilitéCase of 1,000$225.39Supplier Number: 4100OR
- Non sterile tourniquet, white, flat, 1×18"100/box, 10 boxes/CaseLog in pour la disponibilitéCase of 1,000$225.39Supplier Number: 4100W