ContactUsLinkComponent
Cell Stimulation Cocktail, Tonbo Biosciences
Catalogue # 10208-548
Fournisseur:  Tonbo Biosciences
Numéro CAS:  
undefined
Cell Stimulation Cocktail, Tonbo Biosciences
Catalogue # 10208-548
Fournisseur:  Tonbo Biosciences
Supplier Number:  TNB-4975-UL100
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    500X Cell Stimulation Cocktail
  • Size:
    100 µL
  • Cat. no.:
    10208-548
  • Supplier No.:
    TNB-4975-UL100
Spécifications
À propos de cet article
Cell stimulation cocktail is a ready to use cocktail that can be used for polyclonal cell activation. It is composed of the phorbol ester, PMA (Phorbol 12-Myristate 13-Acetate), a calcium ionophore (Ionomycin), and the protein transport inhibitors Brefeldin A and Monensin. It can be used to elicit cytokine production and retention in a variety of cell types allowing for detection of intracellular protein by flow cytometry.
  • Should be stored at –20 °C
  • To be protected from light and tightly sealed to prevent evaporation
It has been quality-tested by treatment of mouse splenocytes for 6 to 18 hours followed by intracellular staining and flow cytometric analysis of IL-2 or Ifn-gamma. It should be used at 1X final concentration for cell stimulation. It is recommended to add the cocktail directly to the cell suspension at 2 uL/mL to obtain a 1X final concentration.
Application: Flow cytometry and functional assays.
Delivery: Supplied as 500X solution in ethanol.