Industrial Gloves, MaxiFlex® Ultimate™ 34-874, Protective Industrial Products
Supplier: PIP
Certificates
About this item
The MaxiFlex® Ultimate™ has become the benchmark for precision handling in dry environments.
- Soft liner made from Nylon and Spandex
- Micro-foam NBR (Nitrile Butadiene Rubber) palm coating for controlled grip
- Silicone free
- FDA compliant, Food contact approved (EU)
- Touchscreen compatible
The patented micro foam nitrile coating offers 360° breathability, reduces hand fatigue and increases comfort. Gloves are washable.
MaxiFlex® has outstanding durability withstanding 18.000 abrasive cycles per millimetre thanks to the use of the DURAtech® coating technology in the coating construction.
Typical applications include: General assembly, micro-engineering, automotive components, painting, and horticulture.
Specifications
- Catalog No:
- 20002-358
- 20002-356
- 20002-354
- 20002-352
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Nylon/Spandex
- Finish:
- Palm coating
- Palm coating
- Palm coating
- Palm coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- FDA regulated:
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +