Corning® Cell Counter, Corning
Corning Cell Counter
76200-994
: Corning
: 6749
This cell counter uses online image processing with a cloud-based machine learning algorithm.
- Fast and accurate results: Fits into the palm of a hand
- Cell count in less than 3 seconds
- Sophisticated deep neural network for cell detection with optimal accuracy – when Trypan blue is added, the system can also detect cell viability
Connect to a Windows® 10 computer or tablet and start the Cytosmart Cloud app. Place the counting chamber on stage, focus on the cells, and press the counting function.
The simplicity of the device allows anyone working in the lab to easily count cells without the need for extensive training. This system utilizes a reusable counting chamber which is included, or a customer supplied hemocytometer.
: CE marked. usTUV, CSA certified.
: Includes a reusable counting chamber.
- Désignation:Corning Cell Counter
- Computer requirements:Windows 10 tablet or PC with a USB 3.0 port
- Cat. No.:76200-994
- Sample volume:10 µL
- W×D×H:122 × 122 × 125 mm
- Field of view:2.0 × 1.5 mm
- Cell size:10 - 70 µm
- Concentration:5 × 10⁴ to 1.0 × 10⁷ cells/ml
- Measurement time:~3 seconds with 73 Mbps download speed and 20 Mbps upload speed
- Magnification:200x
- Weight:1.0 kg
- Resolution:2048 × 1536 Image, Pixels