VWR® ESD Micro Denier Scrub Pant
VWR® ESD Micro Denier Scrub Pant
Supplier:   VWR International
VWR® ESD Micro Denier Scrub Pant
Supplier:  VWR International
Certificates
VWR® ESD Micro Denier Scrub Pant
9 Options Available Below
Product Details & Documents
This style of Scrub Pant is widely used in pharmaceutical cleanroom operations, and meets/exceeds the recommendations outlined in the IEST-RP-CC-003.4, Garment Considerations for Cleanrooms and Other Controlled Environments.
  • Combination elastic back and drawstring front for secure comfort
  • Leg opening with elastic back and three-position snaps for closure
  • Double-needle felled outer seams for extra durability
  • Garments include color-coded loops for easy size identification
  • All edges are pre-serged or laser cut for lint limitation
These scrubs provide an excellent base-layer of protection for any cleanroom garment system. Made with Sandwash material, this lightweight micro-denier fabric is woven from 100% continuous-filament polyester to eliminate any source for lint.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76202-680
  • 76202-682
  • 76202-684
  • 76202-686
  • 76202-688
  • 76202-690
  • 76202-692
  • 76202-696
  • 76202-698
  • 76202-700
  • Garment application:
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Cleanroom|Pharmaceutical
  • Risk area:
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • Gender:
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Fabric:
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
  • 100% continuous-filament polyester
Spécifications
Product Family Options
Informations sur le produit
  • Color
    Taille de confection
    Availability
    Price
  • Navy
    XS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $83.87
    $83.87
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $4,193.28
    $4,193.28
    Prix promotionnel pour les nouveaux clients
    Each
       $83.87
    Case of 50
       $4,193.28
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-680
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $83.87
    $83.87
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $4,193.28
    $4,193.28
    Prix promotionnel pour les nouveaux clients
    Each
       $83.87
    Case of 50
       $4,193.28
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-682
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    M
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $83.87
    $83.87
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $4,193.28
    $4,193.28
    Prix promotionnel pour les nouveaux clients
    Each
       $83.87
    Case of 50
       $4,193.28
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-684
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    L
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $83.87
    $83.87
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $4,193.28
    $4,193.28
    Prix promotionnel pour les nouveaux clients
    Each
       $83.87
    Case of 50
       $4,193.28
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-686
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    XL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $83.87
    $83.87
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $4,193.28
    $4,193.28
    Prix promotionnel pour les nouveaux clients
    Each
       $83.87
    Case of 50
       $4,193.28
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-688
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    XXL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.55
    $87.55
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $4,377.60
    $4,377.60
    Prix promotionnel pour les nouveaux clients
    Each
       $87.55
    Case of 50
       $4,377.60
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-690
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    3XL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $91.11
    $91.11
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $4,555.68
    $4,555.68
    Prix promotionnel pour les nouveaux clients
    Each
       $91.11
    Case of 50
       $4,555.68
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-692
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    4XL
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-696
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    5XL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $95.11
    $95.11
    Prix promotionnel pour les nouveaux clients
    Each
       $95.11
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-698
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester
  • Navy
    6XL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $100.57
    $100.57
    Prix promotionnel pour les nouveaux clients
    Each
       $100.57
    Spécifications:
    More
    Specifications
    Cat. No.
    76202-700
    Garment application
    Cleanroom|Pharmaceutical
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    100% continuous-filament polyester