Sharps Containers, Stackable, Medegen
Catalogue # 89166-424
Fournisseur: Medegen Medical Products, LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Capacity:11.35 L (3 gal.)
- Color:Red
- Description:Sharps Container with Slide Lid
- Cat. no.:89166-424
- Supplier No.:8710
Spécifications
À propos de cet article
Large multi-purpose sharps containers are suitable for OR and dialysis units, and contain locks for final disposal.
Containers feature a tortuous path lid with large and small openings for infectious waste of various types.