6-keto Prostaglandin F1α ELISA Kit, Cayman Chemical Company
Fournisseur: Cayman Chemical Company
Certificats
À propos de cet article
For the quantification of 6-keto PGF1α in various samples.
- Detect 6-keto PGF1α, a surrogate for prostacyclin synthesis in multiple sample types
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure 6-keto PGF1α levels down to 6 pg/mL
- Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
Prostacyclin is formed from arachidonic acid primarily by the vascular endothelium and renal cortex. It is a potent vasodilator and inhibitor of platelet aggregation. PGI2 is non-enzymatically hydrated to 6-keto PGF1α (t½ = 2 – 3 minutes), and then quickly converted to the major metabolite, 2,3-dinor-6-keto PGF1α (t½ = 30 minutes).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Target protein:6-keto-PGF1α
- Mid point:20 – 60 pg/mL
- Detection range:1.6 – 1000 pg/mL
- Sensitivity:6 pg/mL