Prostaglandin F2α ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
Certificates
About this item
For the quantification of PGF2α in tissue culture supernatants and other sample matrices.
- Measure PGF2α in multiple sample types
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure PGF2α levels down to 10 pg/mL
- Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
PGF2α is one of the five primary PGs derived enzymatically directly from the endoperoxide PGH2. The majority of the functional roles ascribed to it relate to fertility, pregnancy, and parturition. Like all of the primary PGs, PGF2α has a very short half-life in the general circulation. The plasma concentration of PGF2α in humans is <10 pg/mL, and probably no more than 1 – 2 pg/mL. Cayman’s PGF2α ELISA Kit is a competitive assay that can be used for the quantification of PGF2α in tissue culture supernatants and other sample matrices.
Specifications
- Assay Type:Competitive
- Mid point:40 – 60 pg/mL
- Detection range:3.9 – 500 pg/mL
- Sensitivity:10 pg/mL