Order Entry
Canada
ContactUsLinkComponent
Anti-SUMO2 Mouse Monoclonal Antibody (CF640R) [clone: SUMO2/1199]
Fournisseur:  Biotium
Certificats
Anti-SUMO2 Mouse Monoclonal Antibody (CF640R) [clone: SUMO2/1199]
Fournisseur:  Biotium

Spécifications

  • Catalog No:
  • 75975-818
  • 75975-816
  • Antigen symbol:
  • SUMO2
  • SUMO2
  • Antigen name:
  • SUMO-2
  • SUMO-2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • SUMO2/1199
  • SUMO2/1199
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 474005
  • Unigene ID: 474005
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Spécifications

Frequently Bought Together