Order Entry
Canada
ContactUsLinkComponent
Anti-PTPRC Mouse Monoclonal Antibody (CF647) [clone: PTPRC/818]
Anti-PTPRC Mouse Monoclonal Antibody (CF647) [clone: PTPRC/818]
Supplier:  Biotium
Certificates
Anti-PTPRC Mouse Monoclonal Antibody (CF647) [clone: PTPRC/818]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75911-546
  • 75911-544
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • Antigen name:
  • CD45RA
  • CD45RA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • PTPRC/818
  • PTPRC/818
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 654514
  • Unigene ID: 654514
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together