Order Entry
Canada
ContactUsLinkComponent
Anti-SUMO2 & SUMO3 Mouse Monoclonal Antibody (CF594) [clone: SM23/496]
Fournisseur:  Biotium
Certificats
Anti-SUMO2 & SUMO3 Mouse Monoclonal Antibody (CF594) [clone: SM23/496]
Fournisseur:  Biotium

Spécifications

  • Catalog No:
  • 75974-600
  • 75974-602
  • Antigen symbol:
  • SUMO2 & SUMO3
  • SUMO2 & SUMO3
  • Antigen name:
  • SUMO-2/3
  • SUMO-2/3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • SM23/496
  • SM23/496
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Spécifications

Frequently Bought Together