Anti-SLC3A2 Mouse Monoclonal Antibody [clone: UM7F8]
Catalogue # 75908-962
Fournisseur: Biotium
Spécifications
- Antibody type:Primary
- Antigen name:CD98
- Antigen symbol:SLC3A2
- Clonality:Monoclonal
- Clone:UM7F8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:50 μL
- Gene ID:Unigene ID: 502769
- Storage buffer:PBS, no BSA, no azide
- Concentration:1 mg/mL
- Cat. no.:75908-962
- Supplier No.:BNUM0425-50
Spécifications
À propos de cet article
CD98 exits as a heterodimer containing a disulphide-linked glycosylated heavy chain and a non-glycosylated light chain. It is a member of the solute carrier family and encodes a cell surface, transmembrane protein. The protein exists as the heavy chain of a heterodimer, covalently bound through disulfide bonds to one of several possible light chains. The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids. Alternatively spliced transcript variants, encoding different isoforms, have been characterized.
Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: Clone UM7F8
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human