Lab Scoops | Avantor
Lab Scoops
Supplier:   TWD Scientific
Lab Scoops
Supplier:  TWD Scientific
Certificates
Lab Scoops
2 Options Available Below
Product Details & Documents
Disposable Lab Scoops allow for easy and rapid transfer of milligram quantities. Scoops are available in either anti-static polypropylene or stainless steel construction.
  • Cost effective
  • Reduces time and labor of repetitive weighing
  • Easy dispense packages
These cost-effective scoops are great for rapid transfer of milligram quanities.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Capacité
    Availability
    Price
  • Lab Scoop
    Stainless steel
    6 - 10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 250
    Case of 250 - $953.15
    $953.15
    Case of 250
       $953.15
    Supplier Number: GSS-03-25
  • Lab Scoop
    PP, anti-static
    6 - 10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,500
    Case of 2,500 - $1,060.13
    $1,060.13
    Case of 2,500
       $1,060.13
    Supplier Number: ASPS-03