Culture Tubes
Low-cost polypropylene tubes are available in the larger sizes and capacities required for detailed analysis. A lip at the top reinforces the rim and facilitates handling.
- Disposable polypropylene test tube
- Extra-long, tall format
- Lip at the top reinforces the rim and facilitates handling
- Autoclavable
Multiple sizes available to meet all your needs.
Frequently Bought Together