ContactUsLinkComponent
Baculovirus Titering Service
Catalog # 77705-774
New Product
Baculovirus Titering Service
Catalog # 77705-774
Supplier Number:  94-901
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    77705-774
Specifications
About this item
Baculovirus titering services to help customers maximize expression and reproducibility.
Accurate titering of recombinant baculovirus is an important step in protein production optimization. A high throughput, flow cytometric assay is utilized to quantitate infectious baculovirus particles produced in insect cell culture, allowing for accurate virus titer determination for multiple conditions in less than 24 hours. The assay detects the cell surface expression of gp64 after a short incubation period allowing for a single round of infection. Virus infected cells are then assessed by flow cytometric analysis for gp64 expression.
Ship 0.5 to 1 ml of virus stock (as little as 200 µl can be titered with advance notice).
Ship sterile aliquots in 2 ml screw cap tubes or equivalent. Ship overnight on cold packs.