Bio-Safe NA Complete Counting Cocktail, RPI
61000-506
: RPI
Small Business Enterprise
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Bio-Safe NA Complete Counting Cocktail
- Size:4 L
- Cat. no.:61000-506
- Supplier No.:111198
Bio-Safe NA provides the highest efficiency for counting organic samples, dry filters and samples treated with tissue solubilizers.
- Bluish tint liquid
- Odorless
Effective and economical cocktail for counting dry wipe tests in radiation safety procedures. Compatible with all plastic scintillation vials.
: Research or further manufacturing use only, not for food or drug use.