DNase I Set | Avantor
DNase I Set
250 U
Catalog # 77001-900
Supplier:   Zymo Research
Fournisseur #:  E1010
DNase I Set
Catalog # 77001-900
Supplier:  Zymo Research
CAS Number:  
DNase I Set
DNase I Set
250 U
CAS Number:  
Price
$211.17
Log in pour la disponibilité
Each
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Zymo research DNase I is a recombinant endonuclease that cleaves single- and double-stranded DNA. It digests DNA more efficiently than other DNase I, making it optimal for sensitive applications like NGS (RNA-seq) and PCR. It is extensively purified, and quality controlled to ensure RNase-free and protease-free.
  • Lyophilized enzyme
  • Used for selectively degrading DNA in the presence of RNA
DNase is suited for applications such as nick translation, production of random fragments, cleavage of genomic DNA for footprinting, removal of DNA template after in vitro transcription and removal of DNA from RNA samples prior to applications such as RT-PCR. It is compatible with all of our RNA kits featuring in-column DNase digestion.
Zymo Research DNase I is lyophilized and supplied with DNA digestion buffer. It dissolves in water within seconds and remains stable for up to 10 freeze-thaw cycles.
Ordering information: Customized, OEM, and bulk orders are available on request.
Delivery information: The set supplied with DNA digestion buffer (4 ml).
Spécifications
Spécifications
  • Taille:
    250 U
  • Cat. No.:
    77001-900
  • Enzyme name:
    DNase I
Spécifications