ContactUsLinkComponent
GENESYS™ Visible and UV-Visible Spectrophotometers, Thermo Scientific
Fournisseur:  Thermo Scientific
Certificats
GENESYS™ Visible and UV-Visible Spectrophotometers, Thermo Scientific
Fournisseur:  Thermo Scientific
Spécifications
  • Catalog No:
  • 76176-738
  • 76176-740
  • 76176-742
  • 76176-744
  • 76176-746
  • 76176-748
  • Model:
  • GENESYS™ 40, Dual Beam
  • GENESYS™ 50, Dual Beam
  • GENESYS™ 140, Dual Beam
  • GENESYS™ 150, Dual Beam
  • GENESYS™ 180, Dual Beam
  • BIOMATE™ 160, Dual Beam
  • Photometric accuracy:
  • ±0.002 A at 0.5 A;
    ±0.004 A at 1.0 A;
    ±0.008 A at 2.0 A
  • ±0.002 A at 0.5 A;
    ±0.004 A at 1.0 A;
    ±0.008 A at 2.0 A
  • ±0.002 A at 0.5 A;
    ±0.004 A at 1.0 A;
    ±0.008 A at 2.0 A
  • ±0.002 A at 0.5 A;
    ±0.004 A at 1.0 A;
    ±0.008 A at 2.0 A
  • ±0.002 A at 0.5 A;
    ±0.004 A at 1.0 A;
    ±0.008 A at 2.0 A
  • ±0.002 A at 0.5 A;
    ±0.004 A at 1.0 A;
    ±0.008 A at 2.0 A
  • Photometric range:
  • –3 to +3.5 A
  • –2 to +3.5 A
  • –3 to +3.5 A
  • –2 to +3.5 A
  • –2 to +3.5 A
  • –2 to +3.5 A
  • Wavelength repeatability:
  • <±0.2 nm
  • <±0.2 nm
  • <±0.2 nm
  • <±0.2 nm
  • <±0.2 nm
  • <±0.2 nm
  • Electrical:
  • 100 - 240 V, 50 - 60 Hz
  • 100 - 240 V, 50 - 60 Hz
  • 100 - 240 V, 50 - 60 Hz
  • 100 - 240 V, 50 - 60 Hz
  • 100 - 240 V, 50 - 60 Hz
  • 100 - 240 V, 50 - 60 Hz
  • Light source:
  • Tungsten
  • Xenon flash
  • Tungsten
  • Xenon flash
  • Xenon flash
  • Xenon flash
  • Repeatability:
  • Photometric: ±0.001 A at 1 A (Measured at 1.0 A at 546 nm)
  • Photometric: ±0.001 A at 1 A (Measured at 1.0 A at 546 nm)
  • Photometric: ±0.001 A at 1 A (Measured at 1.0 A at 546 nm)
  • Photometric: ±0.001 A at 1 A (Measured at 1.0 A at 546 nm)
  • Photometric: ±0.001 A at 1 A (Measured at 1.0 A at 546 nm)
  • Photometric: ±0.001 A at 1 A (Measured at 1.0 A at 546 nm)
  • Stray light:
  • <0.05% T at 340 nm and 400 nm
  • < 1.0% T 198 nm (KCl) , <0.05% T at 220 nm (NaI), <0.03% T at 340 nm (NaNO₂)
  • <0.05% T at 340 nm and 400 nm
  • < 1.0% T 198 nm (KCl) , <0.05% T at 220 nm (NaI), <0.03% T at 340 nm (NaNO₂)
  • < 1.0% T 198 nm (KCl) , <0.05% T at 220 nm (NaI), <0.03% T at 340 nm (NaNO₂)
  • < 1.0% T 198 nm (KCl) , <0.05% T at 220 nm (NaI), <0.03% T at 340 nm (NaNO₂)
  • Dimensions:
  • 355L×385W×195H mm
  • 355L×385W×195H mm
  • 355L×385W×195H mm
  • 355L×385W×195H mm
  • 355L×385W×195H mm
  • 355L×385W×195H mm
  • Weight:
  • 7.5 kg
  • 7.5 kg
  • 7.5 kg
  • 7.5 kg
  • 7.5 kg
  • 7.5 kg